2-Fluorobenzenesulfonyl chloride, 98%
2-Fluorobenzenesulfonyl chloride, 98%
2-Fluorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2-Fluorobenzenesulfonyl chloride, 98%

CAS: 2905-21-7 | C6H4ClFO2S | 194.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14181.06
also known as A14181-06
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.70
Save 14.05 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
93.70
Save 14.05 (15%)
Each
Chemical Identifiers
CAS2905-21-7
IUPAC Name2-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClFO2S
InChI KeyZSZKAQCISWFDCQ-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5350-1.5390 @ 20?C
FormLiquid as melt
Appearance (Color)Clear, colorless to yellow
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only