Sebacoyl chloride, 95%
Sebacoyl chloride, 95%
Sebacoyl chloride, 95%
Thermo Scientific Chemicals

Sebacoyl chloride, 95%

CAS: 111-19-3 | C10H16Cl2O2 | 239.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14226.36
also known as A14226-36
Price (USD)
784.65
Online Exclusive
872.00
Save 87.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
784.65
Online Exclusive
872.00
Save 87.35 (10%)
Each
Chemical Identifiers
CAS111-19-3
IUPAC Namedecanedioyl dichloride
Molecular FormulaC10H16Cl2O2
InChI KeyWMPOZLHMGVKUEJ-UHFFFAOYSA-N
SMILESClC(=O)CCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to yellow to pink to brown
FormLiquid
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Identification (FTIR)Conforms
Refractive Index1.4670-1.4720 @ 20?C
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

Solubility
Miscible with hydrocarbons and ethers.

Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, bases, amines, alcohols and oxidizing agents.
RUO – Research Use Only

General References:

  1. Karoyo, A. H.; Wilson, L. D. Nano-Sized Cyclodextrin-Based Molecularly Imprinted Polymer Adsorbents for Perfluorinated Compounds—A Mini-Review. Nanomaterials 2015, 5 (2), 981-1003.
  2. Rosa, N.; Martins, G. V.; Bastos, M. M. S. M.; Gois, J. R.; Coelho, J. F. J.; Marques, J.; Tavares, C. J.; Magalhaes, F. D. Preparation of robust polyamide microcapsules by interfacial polycondensation of p-phenylenediamine and sebacoyl chloride and plasticization with oleic acid. J. Microencapsulation 2015, 32 (4), 349-357.