3-Fluoroaniline, 98+%
3-Fluoroaniline, 98+%
3-Fluoroaniline, 98+%
3-Fluoroaniline, 98+%
Thermo Scientific Chemicals

3-Fluoroaniline, 98+%

CAS: 372-19-0 | C6H6FN | 111.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14228.36
also known as A14228-36
Price (USD)
719.65
Special offer
Online exclusive
Ends: 30-Jun-2026
846.00
Save 126.35 (15%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
719.65
Special offer
Online exclusive
Ends: 30-Jun-2026
846.00
Save 126.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS372-19-0
IUPAC Name3-fluoroaniline
Molecular FormulaC6H6FN
InChI KeyQZVQQUVWFIZUBQ-UHFFFAOYSA-N
SMILESNC1=CC=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless or yellow to brown
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.

Solubility
Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhou, Y.; Yuan, Y.; You, L.; Anslyn, E. V. Dynamic Aminal-Based TPA Ligands. Chem. Eur. J. 2015, 21 (22), 8207-8213.
  2. Yashkina, E. A.; Yashkin, S. N.; Svetlov, D. A.; Gorshkov, V. V. Themodynamics of adsorption and patterns of the gas-chromatographic retention of cyclic amines on surfaces of graphitized thermal carbon black. Russ. J. Phys. Chem. A 2015, 89 (9), 1672-1682.