Ethyl chlorodifluoroacetate, 98%
Ethyl chlorodifluoroacetate, 98%
Ethyl chlorodifluoroacetate, 98%
Thermo Scientific Chemicals

Ethyl chlorodifluoroacetate, 98%

CAS: 383-62-0 | C4H5ClF2O2 | 158.529 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A14232.22
also known as A14232-22
Price (USD)
183.65
Special offer
Online exclusive
Ends: 30-Jun-2026
216.00
Save 32.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
183.65
Special offer
Online exclusive
Ends: 30-Jun-2026
216.00
Save 32.35 (15%)
Each
Chemical Identifiers
CAS383-62-0
IUPAC Nameethyl 2-chloro-2,2-difluoroacetate
Molecular FormulaC4H5ClF2O2
InChI KeyGVCAWQUJCHZRCB-UHFFFAOYSA-N
SMILESCCOC(=O)C(F)(F)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.3555-1.3595 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only