5-Chloro-2-methoxyaniline, 98%
5-Chloro-2-methoxyaniline, 98%
5-Chloro-2-methoxyaniline, 98%
5-Chloro-2-methoxyaniline, 98%
Thermo Scientific Chemicals

5-Chloro-2-methoxyaniline, 98%

CAS: 95-03-4 | C7H8ClNO | 157.597 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A14244.36
also known as A14244-36
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS95-03-4
IUPAC Name5-chloro-2-methoxyaniline
Molecular FormulaC7H8ClNO
InChI KeyWBSMIPLNPSCJFS-UHFFFAOYSA-N
SMILESCOC1=CC=C(Cl)C=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to dark brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)77.5-83.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only