1-Phenyl-1H-pyrazole, 98%
1-Phenyl-1H-pyrazole, 98%
1-Phenyl-1H-pyrazole, 98%
Thermo Scientific Chemicals

1-Phenyl-1H-pyrazole, 98%

CAS: 1126-00-7 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14270.30
also known as A14270-30
Price (USD)
831.65
Online Exclusive
924.00
Save 92.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
831.65
Online Exclusive
924.00
Save 92.35 (10%)
Each
Chemical Identifiers
CAS1126-00-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
Refractive Index1.5955-1.5995 @ 20?C
Assay (GC)≥97.5%
FormLiquid
1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1H-pyrazole is used in the preparation of 4,5-diphenylpyrazolo[1,5-a]quinoline, 1-(1,2,3,4-tetraphenylnaphthalen-5-yl)pyrazole and 1-(1,2,3,4,5,6,7,8-octaphenylanthracen-9-yl)pyrazole. It acts as a cyclometallated ligand in the preparation of new heteroleptic iridium(III) complexes.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

General References:

  1. Nobuyoshi Umeda; Koji Hirano; Tetsuya Satoh; Naoto Shibata; Hirofumi Sato; Masahiro Miura. Rhodium-catalyzed oxidative 1:1, 1:2, and 1:4 coupling reactions of phenylazoles with internal alkynes through the regioselective cleavages of multiple C-H bonds. Journal of Organic Chemistry.2011, 76 (1), 13-24.
  2. Indira Fabre; Niklas von Wolff; Gaëtan Le Duc; Emmanuel Ferrer Flegeau; Christian Bruneau; Pierre H Dixneuf; Anny Jutand. Autocatalytic intermolecular versus intramolecular deprotonation in C-H bond activation of functionalized arenes by ruthenium(II) or palladium(II) complexes. Chemistry: A European Journal.2013, 19 (23), 7595-7604.