3-Nitrobenzyl bromide, 98+%
3-Nitrobenzyl bromide, 98+%
3-Nitrobenzyl bromide, 98+%
3-Nitrobenzyl bromide, 98+%
Thermo Scientific Chemicals

3-Nitrobenzyl bromide, 98+%

CAS: 3958-57-4 | C7H6BrNO2 | 216.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14310.06
also known as A14310-06
Price (USD)
34.65
Special offer
Online exclusive
Ends: 31-Dec-2025
40.30
Save 5.65 (14%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 31-Dec-2025
40.30
Save 5.65 (14%)
Each
Add to cart
Chemical Identifiers
CAS3958-57-4
IUPAC Name1-(bromomethyl)-3-nitrobenzene
Molecular FormulaC7H6BrNO2
InChI KeyLNWXALCHPJANMJ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(CBr)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)54.0-60.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only