1,9-Dibromononane, 97%
1,9-Dibromononane, 97%
1,9-Dibromononane, 97%
Thermo Scientific Chemicals

1,9-Dibromononane, 97%

CAS: 4549-33-1 | C9H18Br2 | 286.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
Catalog number A14324.09
also known as A14324-09
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.40
Save 7.75 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.40
Save 7.75 (15%)
Each
Chemical Identifiers
CAS4549-33-1
IUPAC Name1,9-dibromononane
Molecular FormulaC9H18Br2
InChI KeyWGAXVZXBFBHLMC-UHFFFAOYSA-N
SMILESBrCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4945-1.4995 @ 20?C
Assay (GC)≥96.0%
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only