3-(3,4-Dimethoxyphenyl)propionic acid, 98%
3-(3,4-Dimethoxyphenyl)propionic acid, 98%
3-(3,4-Dimethoxyphenyl)propionic acid, 98%
Thermo Scientific Chemicals

3-(3,4-Dimethoxyphenyl)propionic acid, 98%

CAS: 2107-70-2 | C11H14O4 | 210.229 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14325.06
also known as A14325-06
Price (USD)
39.65
Online Exclusive
43.70
Save 4.05 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.70
Save 4.05 (9%)
Each
Chemical Identifiers
CAS2107-70-2
IUPAC Name3-(3,4-dimethoxyphenyl)propanoic acid
Molecular FormulaC11H14O4
InChI KeyLHHKQWQTBCTDQM-UHFFFAOYSA-N
SMILESCOC1=CC=C(CCC(O)=O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)94.0-101.0?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only