2',3',4'-Trihydroxyacetophenone, 98%
2',3',4'-Trihydroxyacetophenone, 98%
2',3',4'-Trihydroxyacetophenone, 98%
Thermo Scientific Chemicals

2',3',4'-Trihydroxyacetophenone, 98%

CAS: 528-21-2 | C8H8O4 | 168.148 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14364.06
also known as A14364-06
Price (USD)
44.65
Online Exclusive
49.70
Save 5.05 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.70
Save 5.05 (10%)
Each
Chemical Identifiers
CAS528-21-2
IUPAC Name1-(2,3,4-trihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O4
InChI KeyXIROXSOOOAZHLL-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(O)C(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or pale cream
Melting Point (clear melt)168.5-174.5?C
FormPowder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only