3,5-Dimethoxybenzoic acid, 99%
3,5-Dimethoxybenzoic acid, 99%
3,5-Dimethoxybenzoic acid, 99%
Thermo Scientific Chemicals

3,5-Dimethoxybenzoic acid, 99%

CAS: 1132-21-4 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14374.14
also known as A14374-14
Price (USD)
46.65
Online Exclusive
51.60
Save 4.95 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
51.60
Save 4.95 (10%)
Each
Chemical Identifiers
CAS1132-21-4
IUPAC Name3,5-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyIWPZKOJSYQZABD-UHFFFAOYSA-N
SMILESCOC1=CC(=CC(OC)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)181.0-187.0°C
FormPowder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only