Fluorobenzene, 99%
Fluorobenzene, 99%
Fluorobenzene, 99%
Thermo Scientific Chemicals

Fluorobenzene, 99%

CAS: 462-06-6 | C6H5F | 96.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A14384.30
also known as A14384-30
Price (USD)
68.40
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
68.40
Each
Add to cart
Chemical Identifiers
CAS462-06-6
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.4645-1.4675 @ 20?C
Fluorobenzene is used as a reagent for plastic or resin polymers. It acts as a solvent for highly reactive species. On fluorination of fluorobenzene it gives 1,2-difluorobenzene. It is also used as a material in making medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorobenzene is used as a reagent for plastic or resin polymers. It acts as a solvent for highly reactive species. On fluorination of fluorobenzene it gives 1,2-difluorobenzene. It is also used as a material in making medicine.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
RUO – Research Use Only

General References:

  1. Dine, T. M.; Erb, W.; Berhault, Y.; Rouden, J.; Blanchet, J. Catalytic Chemical Amide Synthesis at Room Temperature: One More Step Toward Peptide Synthesis. J. Org. Chem. 2015, 80 (9), 4532-4544.
  2. Labios, L. A.; Heiden, Z. M.; Mock, M. T. Electronic and Steric Influences of Pendant Amine Groups on the Protonation of Molybdenum Bis(dinitrogen) Complexes. Inorg. Chem. 2015, 54 (9), 4409-4422.