1,3-Difluorobenzene, 99%
1,3-Difluorobenzene, 99%
1,3-Difluorobenzene, 99%
Thermo Scientific Chemicals

1,3-Difluorobenzene, 99%

CAS: 372-18-9 | C6H4F2 | 114.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14387.09
also known as A14387-09
Price (USD)
67.10
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
67.10
Each
Chemical Identifiers
CAS372-18-9
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4365-1.4395 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.

Solubility
Soluble in water at 20°C 1.1g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. M. T. Kirchner, et. al. 1,3-Difluoro­benzene.Acta Cryst.,2009,E652668-2669.
  2. Lise Nygaard, et al.Microwave spectra and dipole moments of 1,2-difluorobenzene and 1,3-difluorobenzene.Spectrochim. Acta, Part A,1967,23(11), 2813-2819.
  3. Lithiation occurs at the 2-position. For a study of lithiation under various conditions, see: J. Chem. Soc., Perkin 1, 2729 (1995).