1-Iodopropane, 98+%, stab. with copper
1-Iodopropane, 98+%, stab. with copper
1-Iodopropane, 98+%, stab. with copper
Thermo Scientific Chemicals

1-Iodopropane, 98+%, stab. with copper

CAS: 107-08-4 | C3H7I | 169.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A14388.36
also known as A14388-36
Price (USD)
162.65
Online Exclusive
181.00
Save 18.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
162.65
Online Exclusive
181.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS107-08-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Identification (FTIR)Conforms
StabilizerCopper powder
Assay (GC)≥98.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Iodopropane is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents etc. It is also used as a solvent and intermediate of organic synthesis.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.
RUO – Research Use Only

General References:

  1. Sang Tae Park.; Sang Kyu Kim.; Myung Soo Kim. Observation of conformation-specific pathways in the photodissociation of 1-iodopropane ions. Nature. 2002, 415 306-308.
  2. Cynthia J. Jenks.; Brian E. Bent.; Neal Bernstein.; Francisco Zaera. Applequist. Chemistry of 1-iodopropane on copper(110): formation, bonding, and reactions of adsorbed propyl groups. J. Am. Chem. Soc. 1993, 115 (1), 308-314.