1-Hexadecanesulfonyl chloride, 97+%
1-Hexadecanesulfonyl chloride, 97+%
1-Hexadecanesulfonyl chloride, 97+%
1-Hexadecanesulfonyl chloride, 97+%
Thermo Scientific Chemicals

1-Hexadecanesulfonyl chloride, 97+%

CAS: 38775-38-1 | C16H33ClO2S | 324.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14396.09
also known as A14396-09
Price (USD)
88.65
Online Exclusive
98.70
Save 10.05 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.70
Save 10.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS38775-38-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Chloride)≥97.50to ≤103.0%
FormPowder
Melting Point (clear melt)54.0-60.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only