Alizarin, 94%
Alizarin, 94%
Alizarin, 94%
Thermo Scientific™

Alizarin, 94%

CAS: 72-48-0 | C14H8O4 | 240.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A14404.22
also known as A14404-22
Price (USD)
55.65
Online Exclusive
61.80
Save 6.15 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.80
Save 6.15 (10%)
Each
Chemical Identifiers
CAS72-48-0
IUPAC Name1,2-dihydroxy-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O4
InChI KeyRGCKGOZRHPZPFP-UHFFFAOYSA-N
SMILESOC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
Water Content (Karl Fischer Titration)≤3%
Melting Point (clear melt)279.0-293.0?C
Assay (HPLC)≥92.0%
FormPowder
View more
Alizarin has a major application in the manufacture of the madder lake pigments which is commonly called as Rose madder and Alizarin crimson. It is involved in the studies of bone growth, osteoporosis, bone marrow and calcium deposits in the vascular system. It is also used as a stain in calcite and aragonite to identify calcium carbonate, and synovial fluid to asses for basic calcium phosphate crystals. It is used in colorimetric measurements for quantification of amine extraction by model food simulants from epoxy polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alizarin has a major application in the manufacture of the madder lake pigments which is commonly called as Rose madder and Alizarin crimson. It is involved in the studies of bone growth, osteoporosis, bone marrow and calcium deposits in the vascular system. It is also used as a stain in calcite and aragonite to identify calcium carbonate, and synovial fluid to asses for basic calcium phosphate crystals. It is used in colorimetric measurements for quantification of amine extraction by model food simulants from epoxy polymer.

Solubility
Soluble in hexane and chloroform. Slightly soluble in water.

Notes
Incompatible with alkalis and oxidizing agents.
RUO – Research Use Only

General References:

  1. Acid-base indicator: pH 5.8 - 7.2, 11.0 - 13.0.
  2. Dai, C.; Song, P.; Wadhawan, J. D.; Fisher, A. C.; Lawerence, N. S. Screen Printed Alizarin-Based Carbon Electrodes: Monitoring pH in Unbuffered Media. Electroanalyis 2015, 27 (4), 917-923.
  3. Amat, A.; Miliani, C.; Romani, A.; Fantacci, S. DFT/TDDFT investigation on the UV-vis absorption and fluorescence properties of alizarin dye. Phys. Chem. 2015, 17, 6374-6382.