2-Nitro-4-(trifluoromethyl)aniline, 98%
2-Nitro-4-(trifluoromethyl)aniline, 98%
2-Nitro-4-(trifluoromethyl)aniline, 98%
2-Nitro-4-(trifluoromethyl)aniline, 98%
Thermo Scientific Chemicals

2-Nitro-4-(trifluoromethyl)aniline, 98%

CAS: 400-98-6 | C7H5F3N2O2 | 206.124 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14413.09
also known as A14413-09
Price (USD)
39.65
Online Exclusive
44.20
Save 4.55 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
44.20
Save 4.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS400-98-6
IUPAC Name2-nitro-4-(trifluoromethyl)aniline
Molecular FormulaC7H5F3N2O2
InChI KeyATXBGHLILIABGX-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)104.0-110.0?C
Appearance (Color)Yellow to brown
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only