Phthalic acid, 99%
Phthalic acid, 99%
Phthalic acid, 99%
Thermo Scientific Chemicals

Phthalic acid, 99%

CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14450.0B
also known as A14450-0B
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Chemical Identifiers
CAS88-99-3
IUPAC Namebenzene-1,2-dicarboxylic acid
Molecular FormulaC8H6O4
InChI KeyXNGIFLGASWRNHJ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Silylated GC)≥98.5%
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

Solubility
Soluble in water, alcohol and etherSoluble in alcohol. Slightly soluble in water, methanol and ether. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. La Nasa, J.; Degano, I.; Modugno, F.; Colombini, M. P. Industrial alkyd resins: characterization of pentaerythritol and phthalic acid esters using integrated mass spectrometry. Rapid Commun. Mass Spectrom. 2015, 29 (3), 225-237.
  2. Tummanapelli, A. K.; Vasudevan, S. Estimating successive pKa values of polyprotic acids from ab initio molecular dynamics using metadynamics: the dissociation of phthalic acid and its isomers. Phys. Chem. Chem. Phys. 2015, 17 (9), 6383-6388.