1-Naphthoyl chloride, 98%
1-Naphthoyl chloride, 98%
1-Naphthoyl chloride, 98%
1-Naphthoyl chloride, 98%
Thermo Scientific Chemicals

1-Naphthoyl chloride, 98%

CAS: 879-18-5 | C11H7ClO | 190.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14460.30
also known as A14460-30
Price (USD)
684.65
Online Exclusive
760.00
Save 75.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
684.65
Online Exclusive
760.00
Save 75.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS879-18-5
IUPAC Namenaphthalene-1-carbonyl chloride
Molecular FormulaC11H7ClO
InChI KeyNSNPSJGHTQIXDO-UHFFFAOYSA-N
SMILESClC(=O)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.6500-1.6540 @ 20?C
Appearance (Color)Clear pale yellow to yellow to pale brown to orange-brown or pale purple
FormLiquid
1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, amines, alcohols, active metals and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Agnieszka Zgoła-Grześkowiak, et al. Solid-phase extraction combined with dispersive liquid-liquid microextraction, fast derivatisation and high performance liquid chromatography-tandem mass spectrometry analysis for trace determination of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol in environmental water samples.J Chromatogr A.,2012,125140-47 .
  2. Vincenzo Lippolis, et al. Improvement of detection sensitivity of T-2 and HT-2 toxins using different fluorescent labeling reagents by high-performance liquid chromatography.Talanta.,2008,74(5), 1476-1483.