2,6-Dimethylbenzonitrile, 97%
2,6-Dimethylbenzonitrile, 97%
2,6-Dimethylbenzonitrile, 97%
Thermo Scientific Chemicals

2,6-Dimethylbenzonitrile, 97%

CAS: 6575-13-9 | C9H9N | 131.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14461.03
also known as A14461-03
Price (USD)
35.65
Online Exclusive
39.10
Save 3.45 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.10
Save 3.45 (9%)
Each
Chemical Identifiers
CAS6575-13-9
IUPAC Name2,6-dimethylbenzonitrile
Molecular FormulaC9H9N
InChI KeyQSACPWSIIRFHHR-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)85.0-94.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only