Allyltriphenylphosphonium bromide, 99%
Allyltriphenylphosphonium bromide, 99%
Allyltriphenylphosphonium bromide, 99%
Thermo Scientific Chemicals

Allyltriphenylphosphonium bromide, 99%

CAS: 1560-54-9 | C21H20BrP | 383.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A14490.30
also known as A14490-30
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
187.00
Save 28.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
187.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS1560-54-9
IUPAC Nametriphenyl(prop-2-en-1-yl)phosphanium bromide
Molecular FormulaC21H20BrP
InChI KeyFWYKRJUVEOBFGH-UHFFFAOYSA-M
SMILES[Br-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Bromide)≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)≤1%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only