3-(Trifluoromethyl)benzoyl chloride, 97%
3-(Trifluoromethyl)benzoyl chloride, 97%
3-(Trifluoromethyl)benzoyl chloride, 97%
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzoyl chloride, 97%

CAS: 2251-65-2 | C8H4ClF3O | 208.564 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14493.06
also known as A14493-06
Price (USD)
56.65
Online Exclusive
63.30
Save 6.65 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
63.30
Save 6.65 (11%)
Each
Chemical Identifiers
CAS2251-65-2
IUPAC Name3-(trifluoromethyl)benzoyl chloride
Molecular FormulaC8H4ClF3O
InChI KeyRUJYJCANMOTJMO-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4755-1.4805 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only