RPMI, no methionine
RPMI, no methionine
RPMI, no methionine
Gibco™

RPMI, no methionine

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as aRead more
Have Questions?
Catalog NumberQuantity
A1451701500 mL
Catalog number A1451701
Price (USD)
174.00
Each
Quantity:
500 mL
Price (USD)
174.00
Each

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 medium has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using our media selector tool.


This RPMI medium is modified as follows:
WithWithout
• L-glutamine• Methionine
• Phenol red• HEPES



Gibco™ RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.

Product Intended Use
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

RPMI 1640 medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 medium requires supplementation, commonly with 10% fetal bovine serum (FBS). RPMI 1640 medium uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Specifications
Amino AcidsNo Methionine
Cell LineHeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell TypeLeukemic Cells
Concentration1 X
For Use With (Application)Mammalian Cell Culture
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeRPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Regulatory StatusRUO
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesGlutamine, Phenol Red, Sodium Bicarbonate
Without AdditivesNo HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8 Deg;C. Protect from light



Shipping conditions: Ambient



Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the density (g/L) for RPMI 1640 Medium?

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer RPMI 1640 medium without both methionine and phenol red?

We offer RPMI 1640 Medium, no methionine (Cat. No. A1451701) and also offer RPMI Medium, no phenol red (Cat. Nos. 11835030, 11835055, 11835063, and 11835105). However, we do not offer RPMI 1640 medium without both methionine and phenol red as a standard catalog product. Our Customs group can make it for you as a custom media. If you are interested, please complete this Custom order inquiry form PDF (https://www.thermofisher.com/content/dam/LifeTech/global/applied-sciences/pdfs/Bioproduction/Gibco-Custom-Media-Buffers-Request-Form.pdf) and email it to custommedia@thermofisher.com. Our Customs group will review your requirements and get back to you within a couple of days with pricing, ordering information, and lead time. After that, you have the choice of proceeding with an “official” order or declining.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.