Potassium L-tartrate hemihydrate, 99%
Potassium L-tartrate hemihydrate, 99%
Potassium L-tartrate hemihydrate, 99%
Thermo Scientific Chemicals

Potassium L-tartrate hemihydrate, 99%

CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14556.0B
also known as A14556-0B
Price (USD)
150.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
150.00
Each
Chemical Identifiers
CAS6100-19-2
IUPAC Namebis(2,3-dihydroxybutanedioic acid) hydrate tetrapotassium
Molecular FormulaC8H14K4O13
InChI KeyDFVWEDQSAWCYPI-UHFFFAOYNA-N
SMILESO.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White
Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only
Paula Vera-Cruz; Rubén A. Toscano; Jorge Balmaseda; Mario Basterrechea; Netzahualcoyotl Niño and Luis Felipe del Castillo. Synthesis and crystal structure of three new cadmium tartrates with open frameworks. CrystEngComm. 2012, 14 (24), 8606-8614.