2-Naphthol, 98+%
2-Naphthol, 98+%
2-Naphthol, 98+%
Thermo Scientific Chemicals

2-Naphthol, 98+%

CAS: 135-19-3 | C10H8O | 144.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14564.0B
also known as A14564-0B
Price (USD)
73.80
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
73.80
Each
Chemical Identifiers
CAS135-19-3
IUPAC Namenaphthalen-2-ol
Molecular FormulaC10H8O
InChI KeyJWAZRIHNYRIHIV-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormFlakes
Appearance (Color)White to cream to pale brown or pale pink
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point (clear melt)118.0-125.0°C
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.

Solubility
Partly miscible in water (0.76 g/L).

Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M. Panizza; P.A.Michaud, G.Cerisola; Ch. Comninellis. Anodic oxidation of 2-naphthol at boron-doped diamond electrodes. Journal of Electroanalytical Chemistry. 2001, 507, (1-2),206-214
  2. William R.Laws; Ludwig. Brand. Analysis of two-state excited-state reactions. The fluorescence decay of 2-naphthol. J. Phys. Chem. 1979, 83, (7),795-802