3-(4-Hydroxyphenyl)propionic acid, 99%
3-(4-Hydroxyphenyl)propionic acid, 99%
3-(4-Hydroxyphenyl)propionic acid, 99%
Thermo Scientific Chemicals

3-(4-Hydroxyphenyl)propionic acid, 99%

CAS: 501-97-3 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14567.30
also known as A14567-30
Price (USD)
414.65
Online Exclusive
460.00
Save 45.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
414.65
Online Exclusive
460.00
Save 45.35 (10%)
Each
Chemical Identifiers
CAS501-97-3
IUPAC Name3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC9H10O3
InChI KeyNMHMNPHRMNGLLB-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder or granules
Melting Point (clear melt)126-133?C
Appearance (Color)White to cream to pale brown
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
View more
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Bakthavatsalam Sundararaju; Alfred A.Antson; Robert S.Phillips; Tatyana V.Demidkina; Maria V.Barbolina; Paul Gollnick ; G.Guy Dodson; Keith S.Wilson. The crystal structure of Citrobacter freundii tyrosine phenol-lyase complexed with 3-(4‘-Hydroxyphenyl)propionic Acid, together with site-directed mutagenesis and kinetic analysis, demonstrates that arginine 381 is required for substrate specificity. Biochemistry. 1997, 36, (21),6502-6510
  2. Hans R.Kricheldorf; Arnd Conradi. New polymer syntheses 16. LC-copolyesters of 3-(4-hydroxyphenyl) propionic acid and 4-hydroxy benzoic acids. Journal of Polymer Science Part A: Polymer Chemistry. 1987, 25, (2),489-504