Ethyl 4-iodobenzoate, 98%
Ethyl 4-iodobenzoate, 98%
Ethyl 4-iodobenzoate, 98%
Thermo Scientific Chemicals

Ethyl 4-iodobenzoate, 98%

CAS: 51934-41-9 | C9H9IO2 | 276.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14570.09
also known as A14570-09
Price (USD)
57.65
Online Exclusive
63.80
Save 6.15 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
63.80
Save 6.15 (10%)
Each
Chemical Identifiers
CAS51934-41-9
IUPAC Nameethyl 4-iodobenzoate
Molecular FormulaC9H9IO2
InChI KeyYCBJOQUNPLTBGG-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5860-1.5900 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only