2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%
2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%
2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%
Thermo Scientific Chemicals

2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%

CAS: 90-24-4 | C10H12O4 | 196.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A14585.06
also known as A14585-06
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS90-24-4
IUPAC Name1-(2-hydroxy-4,6-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O4
InChI KeyFBUBVLUPUDBFME-UHFFFAOYSA-N
SMILESCOC1=CC(O)=C(C(C)=O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Appearance (Color)White to cream
Melting Point79.5-83.5?C
2'-Hydroxy-4',6'-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-4′,6′-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Calixto J.B; Miguel O.G; Yunes R.A; Rae G.A. Action of 2-hydroxy-4,6-dimethoxyacetophenone isolated from Sebastiania schottiana. Planta Medica. 1990, 56, (1),31-35
  2. Marisi G.Soare; Ana P.V. de Felippe; Elsie F.Guimaraes; Massuo J.Kato; Javier Ellena; Antonio C.Doriguetto. 2-Hydroxy-4,6-dimethoxyacetophenone from leaves of Peperomia glabella. J. Braz. Chem. Soc. 2006, 17, (7)