Glutaric acid, 99%
Glutaric acid, 99%
Glutaric acid, 99%
Thermo Scientific Chemicals

Glutaric acid, 99%

CAS: 110-94-1 | C5H8O4 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14595.14
also known as A14595-14
Price (USD)
29.70
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
29.70
Each
Add to cart
Chemical Identifiers
CAS110-94-1
IUPAC Namepentanedioic acid
Molecular FormulaC5H8O4
InChI KeyJFCQEDHGNNZCLN-UHFFFAOYSA-N
SMILESOC(=O)CCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5%
Appearance (Color)White to pale cream
Assay (Silylated GC)≥98.5%
Melting Point95.0-101.0?C
FormCrystals or powder or crystalline powder
View more
Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric acid is used as the raw material for organic synthesis, pharmaceutical intermediate and synthetic resin. It serves as a precursor in the production of polyester polyols, polyamides, ester plasticizers and corrosion inhibitors. It is useful to decrease polymer elasticity and in the synthesis surfactants and metal finishing compounds. It acts as an intermediate during the catabolism of lysine in mammals.

Solubility
Soluble in water, alcohol, benzene and chloroform. Slightly soluble in petroleum ether.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Wang, Y.; Ma, J.; Zhou, Q.; Pang, S.; Zhang, Y. Hygroscopicity of Mixed Glycerol/Mg(NO3)2/Water Droplets Affected by the Interaction between Magnesium Ions and Glycerol Molecules. J. Phys. Chem. B. 2015,119 (17), 5558-5566.
  2. Wang, H.; Li, Z.; Yu, X.; Chen, R.; Xiulai Chen, X.; Liu, L. Green synthesis of (R)-3-TBDMSO glutaric acid methyl monoester using Novozym 435 in non-aqueous media. RSC Adv. 2015, 5 (92), 75160-75166.