4-tert-Butylcatechol, 98%
4-tert-Butylcatechol, 98%
4-tert-Butylcatechol, 98%
4-tert-Butylcatechol, 98%
Thermo Scientific Chemicals

4-tert-Butylcatechol, 98%

CAS: 98-29-3 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14599.30
also known as A14599-30
Price (USD)
56.65
Online Exclusive
62.90
Save 6.25 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.90
Save 6.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS98-29-3
IUPAC Name4-tert-butylbenzene-1,2-diol
Molecular FormulaC10H14O2
InChI KeyXESZUVZBAMCAEJ-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Appearance (Color)White to cream
Identification (FTIR)Conforms
Melting Point (clear melt)52.0-58.0?C
Assay (GC)≥97.5%
4-tert-Butylcatechol is widely utilized as an inhibitor in polymerization of butadiene, styrene, vinyl acetate and other reactive monomers. It plays an important role in the synthesis of tungsten oxide nanoparticles by nonaqueous sol-gel process. It acts as a stabilizer in the manufacturing of polyurethane foam. It is employed as an antioxidant for synthetic rubber, polymers and oil derivatives. It is also utilized as a purification agent for aminoformate catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylcatechol is widely utilized as an inhibitor in polymerization of butadiene, styrene, vinyl acetate and other reactive monomers. It plays an important role in the synthesis of tungsten oxide nanoparticles by nonaqueous sol-gel process. It acts as a stabilizer in the manufacturing of polyurethane foam. It is employed as an antioxidant for synthetic rubber, polymers and oil derivatives. It is also utilized as a purification agent for aminoformate catalysts.

Solubility
Soluble in ether, acetone, alcohol, and methanol. Insoluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Radical inhibitor and antioxidant.
  2. Rafiee, M. Electrochemical oxidation of 4-tert-butylcatechol in the presence of β-cyclodextrin: Interplay between E and CE mechanisms. Int. J. Chem. Kinet. 2012, 44 (8), 507-513.
  3. Nematollahi, D.; Rafiee, M.; Samadi-Maybodi, A. Mechanistic study of electrochemical oxidation of 4-tert-butylcatechol: A facile electrochemical method for the synthesis of new trimer of 4-tert-butylcatechol. Electrochim. Acta 2004, 49 (15), 2495-2502.