Isatoic anhydride, 97%
Isatoic anhydride, 97%
Isatoic anhydride, 97%
Thermo Scientific Chemicals

Isatoic anhydride, 97%

CAS: 118-48-9 | C8H5NO3 | 163.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A14603.36
also known as A14603-36
Price (USD)
105.65
Online Exclusive
117.00
Save 11.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
105.65
Online Exclusive
117.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS118-48-9
IUPAC Name2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Molecular FormulaC8H5NO3
InChI KeyTXJUTRJFNRYTHH-UHFFFAOYSA-N
SMILESO=C1NC2=CC=CC=C2C(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey or cream to pale brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only