2,4,6-Trichlorobenzonitrile, 97+%
Thermo Scientific Chemicals

2,4,6-Trichlorobenzonitrile, 97+%

CAS: 6575-05-9 | C7H2Cl3N | 206.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14609.14
also known as A14609-14
Price (USD)
356.65
Special offer
Online exclusive
Ends: 30-Jun-2026
419.00
Save 62.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
356.65
Special offer
Online exclusive
Ends: 30-Jun-2026
419.00
Save 62.35 (15%)
Each
Chemical Identifiers
CAS6575-05-9
IUPAC Name2,4,6-trichlorobenzonitrile
Molecular FormulaC7H2Cl3N
InChI KeyPGODHCIOIPODFE-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=C(C#N)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay (GC)≥97.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)72.0-82.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only