2,3-Diaminopyridine, 98%
2,3-Diaminopyridine, 98%
2,3-Diaminopyridine, 98%
2,3-Diaminopyridine, 98%
Thermo Scientific Chemicals

2,3-Diaminopyridine, 98%

CAS: 452-58-4 | C5H7N3 | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14618.14
also known as A14618-14
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS452-58-4
IUPAC Namepyridine-2,3-diamine
Molecular FormulaC5H7N3
InChI KeyZZYXNRREDYWPLN-UHFFFAOYSA-N
SMILESNC1=CC=CN=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to brown to black
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)109.0-117.0?C
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

Solubility
Soluble in water (100 g/L).

Notes
It is sensitive to air. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hoong-Kun Fun; Kasthuri Balasubramani. 2,3-Diamino-pyridinium 4-hydroxy-benzoate. Acta Crystallographica Section E. 2009, 65, (Pt 7),o1496-o1497
  2. Xiangyang Shi; Istvan Banyai; Wojciech G.Lesniak; Mohammad T.Islam; Istvan Orszagh; Peter Balogh; James R.Baker; Lajos P.Balogh. Capillary electrophoresis of polycationic poly(amidoamine) dendrimers. Electrophoresis. 2005, 26, (15),2949-2959