4'-Methoxypropiophenone, 99%
4'-Methoxypropiophenone, 99%
4'-Methoxypropiophenone, 99%
4'-Methoxypropiophenone, 99%
Thermo Scientific Chemicals

4'-Methoxypropiophenone, 99%

CAS: 121-97-1 | C10H12O2 | 164.204 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14619.36
also known as A14619-36
Price (USD)
611.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
611.00
Each
Add to cart
Chemical Identifiers
CAS121-97-1
IUPAC Name1-(4-methoxyphenyl)propan-1-one
Molecular FormulaC10H12O2
InChI KeyZJVAWPKTWVFKHG-UHFFFAOYSA-N
SMILESCCC(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLow melting fused solid or clear liquid as melt
Assay (GC)≥98.5%
Refractive Index1.5450-1.5480 as melt
Appearance (Color)Clear colorless to yellow or orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only