Thiosemicarbazide, 98%
Thiosemicarbazide, 98%
Thiosemicarbazide, 98%
Thermo Scientific Chemicals

Thiosemicarbazide, 98%

CAS: 79-19-6 | CH5N3S | 91.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A14630.36
also known as A14630-36
Price (USD)
108.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
108.00
Each
Chemical Identifiers
CAS79-19-6
IUPAC Nameaminothiourea
Molecular FormulaCH5N3S
InChI KeyBRWIZMBXBAOCCF-UHFFFAOYSA-N
SMILESNNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Appearance (Color)White to cream to yellow or pale pink
FormCrystals or powder or crystalline powder
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in TLC to stain alpha-keto acidsThiosemicarbazide is used as a reagent for ketones as well as chemical intermediate. It is involved in the detection of metals. It is also used in photography. Further, it is effective for controlling bacterial leaf blight in rice. In addition to this, it is used to prepare 1,3,4-thiadiazoles.

Solubility
Soluble in water, ethanol, alcohols and hot methanol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

General References:

  1. Hong, L. R.; Wang, J. P.; Zhuo, Y.; Chai, Y. Q.; Zhao, M.; Gui, G. F.; Yuan, R. A regenerable electrochemiluminescence aptasensor incorporating poly(ethylenimine) and thiosemicarbazide as dual co-reactants for signal amplification. Electrochim. Acta 2015, 186, 174-181.
  2. Karthik, N.; Sethuraman, M. G. Improved copper corrosion resistance of epoxy-functionalized hybrid sol-gel monolayers by thiosemicarbazide. Ionics 2015, 21 (5), 1477-1488.