Thiophene-2-carbonyl chloride, 98%
Thiophene-2-carbonyl chloride, 98%
Thiophene-2-carbonyl chloride, 98%
Thiophene-2-carbonyl chloride, 98%
Thermo Scientific Chemicals

Thiophene-2-carbonyl chloride, 98%

CAS: 5271-67-0 | C5H3ClOS | 146.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14641.36
also known as A14641-36
Price (USD)
297.65
Online Exclusive
330.00
Save 32.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
297.65
Online Exclusive
330.00
Save 32.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS5271-67-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5880-1.5920 @ 20?C
Appearance (Color)Clear colorless to pale yellow or brown or green
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only