4-Isopropylaniline, 99%
4-Isopropylaniline, 99%
4-Isopropylaniline, 99%
4-Isopropylaniline, 99%
Thermo Scientific Chemicals

4-Isopropylaniline, 99%

CAS: 99-88-7 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A14646.14
also known as A14646-14
Price (USD)
56.65
Online Exclusive
63.10
Save 6.45 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
63.10
Save 6.45 (10%)
Each
Add to cart
Chemical Identifiers
CAS99-88-7
IUPAC Name4-(propan-2-yl)aniline
Molecular FormulaC9H13N
InChI KeyLRTFPLFDLJYEKT-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to orange to red to brown
Assay (GC)≥98.5%
Refractive Index1.5415-1.5445 @ 20°C (non-U.S. specification)
FormLiquid
Identification (FTIR)Conforms (non-U.S. specification)
4-Isopropylaniline was used in the hydroamination of phenylacetylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropylaniline was used in the hydroamination of phenylacetylene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong acids. Light sensitive
RUO – Research Use Only
S Breitenlechner.; M Fleck.; TE Müller. Solid catalysts on the basis of supported ionic liquids and their use in hydroamination reactions. Journal of Molecular Catalysis A: Chemical.2004214 (1), 175-179.