2-Fluoro-5-nitrotoluene, 98+%
2-Fluoro-5-nitrotoluene, 98+%
2-Fluoro-5-nitrotoluene, 98+%
2-Fluoro-5-nitrotoluene, 98+%
Thermo Scientific Chemicals

2-Fluoro-5-nitrotoluene, 98+%

CAS: 455-88-9 | C7H6FNO2 | 155.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14649.18
also known as A14649-18
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
187.00
Save 28.35 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
158.65
Special offer
Online exclusive
Ends: 30-Jun-2026
187.00
Save 28.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS455-88-9
IUPAC Name1-fluoro-2-methyl-4-nitrobenzene
Molecular FormulaC7H6FNO2
InChI KeyXUCYJGMIICONES-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.0%
Melting Point (clear melt)36.0-42.0?C
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only