1-Hepten-3-ol, 98%
1-Hepten-3-ol, 98%
1-Hepten-3-ol, 98%
Thermo Scientific Chemicals

1-Hepten-3-ol, 98%

CAS: 4938-52-7 | C7H14O | 114.188 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14651.18
also known as A14651-18
Price (USD)
179.65
Special offer
Online exclusive
Ends: 30-Jun-2026
211.00
Save 31.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
179.65
Special offer
Online exclusive
Ends: 30-Jun-2026
211.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS4938-52-7
IUPAC Namehept-1-en-3-ol
Molecular FormulaC7H14O
InChI KeyPZKFYTOLVRCMOA-UHFFFAOYNA-N
SMILESCCCCC(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.4315-1.4355 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only