2,4-Dimethylbenzaldehyde, 90+%
2,4-Dimethylbenzaldehyde, 90+%
2,4-Dimethylbenzaldehyde, 90+%
Thermo Scientific Chemicals

2,4-Dimethylbenzaldehyde, 90+%

CAS: 15764-16-6 | C9H10O | 134.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14655.14
also known as A14655-14
Price (USD)
151.65
Online Exclusive
168.00
Save 16.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
151.65
Online Exclusive
168.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS15764-16-6
IUPAC Name2,4-dimethylbenzaldehyde
Molecular FormulaC9H10O
InChI KeyGISVICWQYMUPJF-UHFFFAOYSA-N
SMILESCC1=CC=C(C=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥90.0%
Free acid (titration)≤5%
Refractive Index1.5095-1.5895 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only