n-Tetradecane, 99%
n-Tetradecane, 99%
n-Tetradecane, 99%
Thermo Scientific Chemicals

n-Tetradecane, 99%

CAS: 629-59-4 | C14H30 | 198.394 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1500 g
2500 g
Catalog number A14672.22
also known as A14672-22
Price (USD)
75.10
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
75.10
Each
Chemical Identifiers
CAS629-59-4
IUPAC Nametetradecane
Molecular FormulaC14H30
InChI KeyBGHCVCJVXZWKCC-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Assay (GC)≥98.5%
FormLiquid
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Brown, P. S.; Bhushan, B. Mechanically durable, superomniphobic coatings prepared by layer-by-layer technique for self-cleaning and anti-smudge. J. Colloid Interface Sci. 2015, 456, 210-218.
  2. Fan, D.; Zheng, P.; Ma, Y.; Yin, T.; Zhao, J.; Shen, W. Effects of water content and chain length of n-alkane on the interaction enthalpy between the droplets in water/sodium bis (2-ethylhexyl)-sulfosuccinate-alkane microemulsions. Soft Matter 2015, 11 (14), 2885-2892.