1-Bromo-4-n-octylbenzene, 97%
1-Bromo-4-n-octylbenzene, 97%
1-Bromo-4-n-octylbenzene, 97%
Thermo Scientific Chemicals

1-Bromo-4-n-octylbenzene, 97%

CAS: 51554-93-9 | C14H21Br | 269.226 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A14676.03
also known as A14676-03
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
55.30
Save 7.65 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
55.30
Save 7.65 (14%)
Each
Chemical Identifiers
CAS51554-93-9
IUPAC Name1-bromo-4-octylbenzene
Molecular FormulaC14H21Br
InChI KeyOOZQSVXPBCINJF-UHFFFAOYSA-N
SMILESCCCCCCCCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Refractive Index1.5120-1.5170 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only