4-(p-Tolyl)butyric acid, 97%
4-(p-Tolyl)butyric acid, 97%
4-(p-Tolyl)butyric acid, 97%
Thermo Scientific Chemicals

4-(p-Tolyl)butyric acid, 97%

CAS: 4521-22-6 | C11H14O2 | 178.231 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14680.14
also known as A14680-14
Price (USD)
680.65
Online Exclusive
756.00
Save 75.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
680.65
Online Exclusive
756.00
Save 75.35 (10%)
Each
Chemical Identifiers
CAS4521-22-6
IUPAC Name4-(4-methylphenyl)butanoic acid
Molecular FormulaC11H14O2
InChI KeyIXWOVMRDYFFXGI-UHFFFAOYSA-N
SMILESCC1=CC=C(CCCC(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)54.0-63.0?C
Appearance (Color)White to cream
Assay (Silylated GC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only