1,1,1-Trifluoro-2,4-pentanedione, 98%
1,1,1-Trifluoro-2,4-pentanedione, 98%
1,1,1-Trifluoro-2,4-pentanedione, 98%
1,1,1-Trifluoro-2,4-pentanedione, 98%
Thermo Scientific Chemicals

1,1,1-Trifluoro-2,4-pentanedione, 98%

CAS: 367-57-7 | C5H5F3O2 | 154.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14682.14
also known as A14682-14
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
107.65
Online Exclusive
119.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS367-57-7
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3855-1.3900 @ 20?C
Appearance (Color)Clear colorless to yellow
CommentMaterial sourced in UK and US
FormLiquid
Assay (GC)≥97.5% (sum of tautomers)
View more
1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.

Solubility
Soluble in water (slightly) and many organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Belen Zarranz; Andres Jaso; Ignacio Aldana; Antonio Monge. Synthesis and anticancer activity evaluation of new 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethyl-quinoxaline 1,4-di-N-oxide derivatives. Bioorganic & Medicinal Chemistry. 2004, 12, (13),3711-3721 
  2. Mansoureh Zahedi-Tabrizi; Fariba Tayyari; Zainab Moosavi-Tekyeh; Alireza Jalali; Sayyed Faramarz Tayyari. Structure and vibrational assignment of the enol form of 1,1,1-trifluoro-2,4-pentanedione. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2006, 65, (2),387-396
  3. Chelating ligand. Review of chemistry of fluorinated ß-diketones: Russ. Chem. Rev., 50, 180 (1981).
  4. For use in the preparation of trifluoromethyl pyrimidines by reaction with guanidines, see: Synth. Commun., 20, 913 (1990):