1,3-Dibromopropane, 98%
1,3-Dibromopropane, 98%
1,3-Dibromopropane, 98%
1,3-Dibromopropane, 98%
Thermo Scientific Chemicals

1,3-Dibromopropane, 98%

1,3-Dibromopropane, 98%, C3H6Br2, CAS Number-109-64-8, 1,3-dibromo propane, 1,3-dibromo-propane, .alpha.,.gamma.-dibromopropane, alpha,gamma-dibromopropane, ccris 6711, .omega.,.omega.'-dibromopropane, propane, 1,3-dibromo, trimethylene bromide, trimethylene dibromide, unii-yqr3048ix9, 5000g, 635662 | CAS: 109-64-8 | C3H6Br2 | 201.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14687.30
also known as A14687-30
Price (USD)
73.65
Online Exclusive
81.80
Save 8.15 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
73.65
Online Exclusive
81.80
Save 8.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS109-64-8
IUPAC Name1,3-dibromopropane
Molecular FormulaC3H6Br2
InChI KeyVEFLKXRACNJHOV-UHFFFAOYSA-N
SMILESBrCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5210-1.5250 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromopropane is used to prepare C3-bridged compounds through C-N coupling reactions. It is used in the preparation of 2,3,4,5-tetrahydro-7,7-diphenylimidazo- [2,1-b]-thiazine-6(7H)-one by reacting with potassium salt of 5,5-diphenyl-2-thiohydantoin.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Ren, C.; Wang, F.; Zhang, Z.; Nie, H.; Li, N.; Cui, M. Synthesis, surface activity and aggregation behavior of Gemini imidazolium surfactants 1,3-bis(3-alkylimidazolium-1-yl) propane bromide. Colloids Surf., A 2015, 467, 1-8.
  2. Kolakowski, R. V.; Young, T. D.; Howard, P. W.; Jeffrey, S. C.; Senter, P. D. Synthesis of a C2-aryl-pyrrolo[2,1-c][1,4]benzodiazepine monomer enabling the convergent construction of symmetrical and non-symmetrical dimeric analogs. Tetrahedron Lett. 2015, 56 (30), 4512-4515.