4-Methylbiphenyl, 98%
4-Methylbiphenyl, 98%
4-Methylbiphenyl, 98%
4-Methylbiphenyl, 98%
Thermo Scientific Chemicals

4-Methylbiphenyl, 98%

CAS: 644-08-6 | C13H12 | 168.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14702.03
also known as A14702-03
Price (USD)
34.65
Online Exclusive
38.60
Save 3.95 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.65
Online Exclusive
38.60
Save 3.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS644-08-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5% (U.K. specification)
Melting Point (clear melt)43.5-49.5°C (U.K. specification)
Appearance (Color)White to cream or pale yellow to pale brown
Assay from Supplier's CofA≥97.5% (GC) (U.S. specification)
FormCrystals or powder or crystalline powder or fused/lumpy solid
View more
4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

Solubility
Insoluble in water. Soluble in alcohol and ether, very soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. D Goubet.; P Meric.; JR Dormoy. New practical syntheses of 4'-methylbiphenyl-2-carbonitrile and-2-carbaldehyde. J. Org. Chem.199964 (12), 4516-4518.
  2. JP Shen.; L Sun.; C Song. Shape-selective synthesis of 4, 4'-dimethylbiphenyl. 1. Methylation of 4-methylbiphenyl over modified zeolite catalysts. Catalysis letters.200065 (1), 147-151.