8-Hydroxyquinoline, 99%
8-Hydroxyquinoline, 99%
8-Hydroxyquinoline, 99%
8-Hydroxyquinoline, 99%
Thermo Scientific Chemicals

8-Hydroxyquinoline, 99%

CAS: 148-24-3 | C9H7NO | 145.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A14720.22
also known as A14720-22
Price (USD)
46.40
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
46.40
Each
Add to cart
Chemical Identifiers
CAS148-24-3
IUPAC Namequinolin-8-ol
Molecular FormulaC9H7NO
InChI KeyMCJGNVYPOGVAJF-UHFFFAOYSA-N
SMILESOC1=C2N=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)71.0-77.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
8-Hydroxyquinoline is used as an antiseptic and disinfectant. Its alcoholic solution is used in liquid bandages. It is used to prepare tris(8-hydroxyquinolinato)aluminum, which finds application as a component of organic light-emitting diodes (OLED′s). It is also used as a reagent and as a stabilizer of hydrogen peroxide in a rocket fuel oxidizer. It is used as a chelating agent and used as a carrier for radio-indium for diagnostic purposes. Its halogenated derivatives are used as anti-infective agents and oral antiamebics.

Solubility
Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene.Soluble in acetic acid, ethanol, acetone, chloroform, benzene and aqueous mineral acids. Insoluble in water and diethylether.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Chelating ligand.
  2. Thinnes, C. C.; Tumber, A.; Yapp, C.; Scozzafava, G.; Yeh, T.; Chan, M. C.; Tran, T. A.; Hsu. K.; Tarhonskaya, H.; Walport, L. J.; Wilkins, S. E.; Martinez, E. D.; Müller, S.; Pugh, C. W.; Ratcliffe, P. J.; Brennan, P. E.; Kawamura, A.; Schofield, C. J. Betti reaction enables efficient synthesis of 8-hydroxyquinoline inhibitors of 2-oxoglutarate oxygenases. Chem. Commun. 2015, 51 (84), 15458-15461.
  3. Liang, S. H.; Southon, A. G.; Fraser, B. H.; Krause-Heuer, A. M.; Zhang, B.; Shoup, T. M.; Lewis, R.; Volitakis, I.; Han, Y.; Greguric, I.; Bush, A. I.; Vasdev, N. Novel fluorinated 8-hydroxyquinoline based metal ionophores for exploring the metal hypothesis of Alzheimer’s disease. ACS Med. Chem. Lett. 2015, 6 (9), 1025-1029.