4-Bromo-o-xylene, 97%
4-Bromo-o-xylene, 97%
4-Bromo-o-xylene, 97%
Thermo Scientific Chemicals

4-Bromo-o-xylene, 97%

CAS: 583-71-1 | C8H9Br | 185.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14736.14
also known as A14736-14
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 28-Dec-2026
109.00
Save 16.35
Each
Chemical Identifiers
CAS583-71-1
IUPAC Name4-bromo-1,2-dimethylbenzene
Molecular FormulaC8H9Br
InChI KeyQOGHRLGTXVMRLM-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5535-1.5585 @ 20?C
Assay (GC)≥96.0%
FormLiquid
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M.Arivazhagan; R.Meenakshi. Vibrational spectroscopic studies and DFT calculations of 4-bromo-o-xylene. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2012, 91, 419-430
  2. G.A.Bagirzade. Kinetics of oxidative ammonolysis of 4-bromo-o-xylene: I. Transformations of 4-bromo-o-xylene and 4-bromo-o-tolunitrile. Russian Journal of General Chemistry. 2010, 80, (8),1672-1676