1-Formylpiperazine, tech. 90
1-Formylpiperazine, tech. 90
1-Formylpiperazine, tech. 90
Thermo Scientific Chemicals

1-Formylpiperazine, tech. 90

CAS: 7755-92-2 | C5H11N2O | 115.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14757.22
also known as A14757-22
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS7755-92-2
IUPAC Name4-formylpiperazin-1-ium
Molecular FormulaC5H11N2O
InChI KeyMSSDTZLYNMFTKN-UHFFFAOYSA-O
SMILESO=CN1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥88.0%
FormLiquid
Refractive Index1.4715-1.5515 @ 20°C
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only