2-Methyl-5-nitrobenzenesulfonyl chloride, 97%
2-Methyl-5-nitrobenzenesulfonyl chloride, 97%
2-Methyl-5-nitrobenzenesulfonyl chloride, 97%
2-Methyl-5-nitrobenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

2-Methyl-5-nitrobenzenesulfonyl chloride, 97%

CAS: 121-02-8 | C7H6ClNO4S | 235.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14760.06
also known as A14760-06
Price (USD)
58.65
Online Exclusive
64.60
Save 5.95 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
64.60
Save 5.95 (9%)
Each
Add to cart
Chemical Identifiers
CAS121-02-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)39.0-49.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only